This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02437399_g1
See other FZR-1 GE Assays ›
Gene Symbol:
fzr-1
Gene Name:
WD_REPEATS_REGION domain-containing protein
Gene Aliases:
CELE_ZK1307.6
Chromosome Location:
Chr.II: 9640156 - 9645233 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02437399_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fzr-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_063674.6 NP_496075.1 4 - 5 1496 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

anaphase-promoting complex-dependent catabolic process
cell cycle G1/S phase transition
regulation of cell division
positive regulation of ubiquitin protein ligase activity
positive regulation of anaphase-promoting complex-dependent catabolic process
anaphase-promoting complex binding
ubiquitin-protein transferase activator activity
ubiquitin ligase activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0