This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02436279_g1
See other D2085.7 GE Assays ›
Gene Symbol:
D2085.7 pyr-1
Gene Name:
Uncharacterized protein
Glutamine-dependent carbamoyl-phosphate synthase
Gene Aliases:
CELE_D2085.7
CELE_D2085.1
Chromosome Location:
Chr.II: 8641381 - 8648232 on Build WBcel235
Chromosome Location:
Chr.II: 8647140 - 8658900 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02436279_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D2085.7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267236.3 NP_001254165.1 1 - 2 98 98
NR_051453.2 4 - 5 3082 98
Gene Symbol: pyr-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_063437.6 NP_495838.2 1 - 2 105 98

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_063437.6 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

nematode larval development
'de novo' pyrimidine nucleobase biosynthetic process
pyrimidine nucleotide biosynthetic process
UTP biosynthetic process
cellular amino acid metabolic process
glutamine metabolic process
nitrogen compound metabolic process
metabolic process
citrulline biosynthetic process
pyrimidine nucleobase biosynthetic process
'de novo' UMP biosynthetic process
pharynx development
pharyngeal gland morphogenesis
nucleotide binding
catalytic activity
aspartate carbamoyltransferase activity
carbamoyl-phosphate synthase (glutamine-hydrolyzing) activity
dihydroorotase activity
ATP binding
amino acid binding
transferase activity
carboxyl- or carbamoyltransferase activity
hydrolase activity
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds
ligase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0