Assay Details
Gene Symbol: cpna-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047258.3 | NP_001040723.1 | 10 - 11 | 7885 | 151 | |
NM_063222.4 | NP_495623.2 | 19 - 20 | 4678 | 151 | ||
NM_063224.6 | NP_495625.3 | 22 - 23 | 22894 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
