Assay Details
Gene Symbol: cpna-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047258.3 | NP_001040723.1 | 4 - 5 | 7192 | 70 | |
NM_063222.4 | NP_495623.2 | 13 - 14 | 3985 | 70 | ||
NM_063224.6 | NP_495625.3 | 16 - 17 | 22201 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
