Assay Details
Gene Symbol: cpna-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047258.3 | NP_001040723.1 | 3 - 4 | 7110 | 71 | |
NM_063222.4 | NP_495623.2 | 12 - 13 | 3903 | 71 | ||
NM_063224.6 | NP_495625.3 | 15 - 16 | 22119 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
