This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02435120_g1
See other SYD-1 GE Assays ›
Gene Symbol:
syd-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F35D2.5
Chromosome Location:
Chr.II: 7585696 - 7594912 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02435120_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: syd-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001026975.4 NP_001022146.1 7 - 8 700 83
NM_001383891.1 5 - 6 520 83

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001383891.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
multicellular organism development
nervous system development
axo-dendritic transport
cell projection organization
cell differentiation
cellular protein localization
locomotion
activation of GTPase activity
GTPase activator activity
receptor binding
Rho GTPase binding
Roundabout binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header