This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02434639_g1
See other GLD-3 GE Assays ›
Gene Symbol:
gld-3
Gene Name:
protein-coding
Gene Aliases:
CELE_T07F8.3
Chromosome Location:
Chr.II: 7132643 - 7137522 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02434639_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gld-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_063078.4 NP_495479.2 4 - 5 386 129
NM_063079.7 NP_495480.1 4 - 5 386 129

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

nuclear division
mitotic cytokinesis
nematode larval development
multicellular organism development
spermatogenesis
embryo development ending in birth or egg hatching
germ-line sex determination
positive regulation of multicellular organism growth
vulval development
masculinization of hermaphroditic germ-line
germ-line stem cell division
RNA polyadenylation
positive regulation of meiotic nuclear division
negative regulation of binding
meiotic cell cycle
RNA binding
polynucleotide adenylyltransferase activity
protein binding
protein domain specific binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0