This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02434401_g1
See other TRA-2 GE Assays ›
Gene Symbol:
tra-2
Gene Name:
protein-coding
Gene Aliases:
CELE_C15F1.3
Chromosome Location:
Chr.II: 6955384 - 6965673 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02434401_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tra-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001381483.1 NP_001367508.1 2 - 3 396 87
NM_063025.9 NP_495426.1 22 - 23 3660 87
GenBank mRNA M91371.1 - 22 - 23 3696 87
S42187.1 - 22 - 23 3696 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
spermatogenesis
sex determination
sex differentiation
female gonad development
germ-line sex determination
cell differentiation
oocyte fate determination
hermaphrodite germ-line sex determination
hermaphrodite somatic sex determination
oocyte development
oocyte morphogenesis
transmembrane signaling receptor activity
protein binding
signaling receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0