This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02432260_g1
See other SMC-5 GE Assays ›
Gene Symbol:
smc-5
Gene Name:
Structural maintenance of chromosomes protein 5
Gene Aliases:
CELE_C27A2.1
Chromosome Location:
Chr.II: 5062125 - 5067129 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02432260_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: smc-5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_062534.5 NP_494935.1 2 - 3 298 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
DNA replication
DNA repair
DNA recombination
cellular response to DNA damage stimulus
sister chromatid cohesion
meiotic DNA double-strand break processing involved in reciprocal meiotic recombination
meiotic cell cycle
nucleotide binding
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0