Assay Details
Gene Symbol: T24E12.10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062245.3 | NP_494646.2 | 24 - 25 | 3106 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: T24E12.10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062245.3 | NP_494646.2 | 24 - 25 | 3106 | 73 |
Set Membership: |
![]() ![]() ![]() ![]() |