This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02429566_m1
See other UBC-15 GE Assays ›
Gene Symbol:
ubc-15
Gene Name:
protein-coding
Gene Aliases:
CELE_Y110A2AR.2
Chromosome Location:
Chr.II: 2900441 - 2913605 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02429566_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ubc-15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001350378.2 NP_001337292.1 2 - 3 331 67
NM_001350379.2 NP_001337293.1 2 - 3 331 67
NM_001350380.2 NP_001337301.1 333 67
NM_001373689.1 NP_001360421.1 2 - 3 331 67
NM_001373690.2 NP_001360422.1 2 - 3 331 67
NM_061996.5 NP_494397.1 2 - 3 333 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001350380.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Process(es)

ubiquitin protein ligase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0