This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02426646_g1
See other ETR-1 GE Assays ›
Gene Symbol:
etr-1
Gene Name:
protein-coding
Gene Aliases:
CELE_T01D1.2
Chromosome Location:
Chr.II: 154166 - 170630 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02426646_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: etr-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001381290.2 NP_001367867.1 5 - 6 841 62
NM_001381291.1 NP_001368521.1 5 - 6 820 62
NM_001381292.2 NP_001367871.1 6 - 7 821 62
NM_001381293.1 NP_001367870.1 6 - 7 932 62
NM_001381294.1 NP_001367872.1 6 - 7 1029 62
NM_001381296.1 NP_001368522.1 4 - 5 407 62
NM_001381297.1 NP_001368523.1 4 - 5 407 62
NM_001381298.1 NP_001368524.1 4 - 5 407 62
NM_001381299.1 NP_001368525.1 2 - 3 281 62
NM_001381300.1 NP_001368526.1 2 - 3 281 62
NM_001381301.1 NP_001368527.1 2 - 3 281 62
NM_001381302.1 NP_001367869.1 1 - 2 83 62
NM_001381303.1 NP_001368528.1 1 - 2 83 62
NM_001381304.1 NP_001368529.1 1 - 2 83 62
NM_001381305.1 NP_001368530.1 1 - 2 83 62
NR_132573.1 5 - 6 779 62
GenBank mRNA EF523766.1 - 5 - 6 779 62
EF523770.1 - 5 - 6 779 62
U53931.1 - 5 - 6 842 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA splice site selection
determination of adult lifespan
nucleic acid binding
RNA binding
mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header