This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02426375_g1
See other POLE-1 GE Assays ›
Gene Symbol:
pole-1
Gene Name:
DNA polymerase epsilon catalytic subunit
Gene Aliases:
CELE_F33H2.5
Chromosome Location:
Chr.I: 15005603 - 15013909 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02426375_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pole-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_061215.5 NP_493616.1 5 - 6 1909 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mitotic cell cycle
DNA replication
leading strand elongation
DNA repair
base-excision repair, gap-filling
nucleotide-excision repair, DNA gap filling
DNA replication proofreading
DNA biosynthetic process
nucleic acid phosphodiester bond hydrolysis
nucleotide binding
nucleic acid binding
DNA binding
DNA-directed DNA polymerase activity
zinc ion binding
single-stranded DNA 3'-5' exodeoxyribonuclease activity
transferase activity
nucleotidyltransferase activity
metal ion binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0