This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02426045_m1
See other HPRT-1 GE Assays ›
Gene Symbol:
hprt-1 Y105E8B.6 riok-2
Gene Name:
Hypoxanthine phosphoribosyltransferase
Uncharacterized protein
Serine/threonine-protein kinase RIO2
Gene Aliases:
CELE_Y105E8B.5
CELE_Y105E8B.6
CELE_Y105E8B.3
Chromosome Location:
Chr.I: 14657355 - 14661495 on Build WBcel235
Chromosome Location:
Chr.I: 14657366 - 14662928 on Build WBcel235
Chromosome Location:
Chr.I: 14652116 - 14661417 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
-
Assay ID Ce02426045_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hprt-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_061144.6 NP_493545.1 1 - 2 79 73
Gene Symbol: Y105E8B.6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001383764.1 1 - 2 68 73
Gene Symbol: riok-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_061143.8 NP_493544.2 3 - 4 1953 73

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001383764.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

purine ribonucleoside salvage
adenine salvage
guanine salvage
nucleoside metabolic process
GMP salvage
IMP salvage
hypoxanthine metabolic process
nematode larval development
protein phosphorylation
phosphorylation
maturation of SSU-rRNA
nucleotide binding
magnesium ion binding
hypoxanthine phosphoribosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
identical protein binding
metal ion binding
guanine phosphoribosyltransferase activity
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0