This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02424853_g1
See other ACOX-1.4 GE Assays ›
Gene Symbol:
acox-1.4
Gene Name:
Acyl-coenzyme A oxidase
Gene Aliases:
CELE_F08A8.4
Chromosome Location:
Chr.I: 12951134 - 12954015 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02424853_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: acox-1.4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_060863.5 NP_493264.1 1 - 2 116 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
fatty acid beta-oxidation
fatty acid beta-oxidation using acyl-CoA oxidase
lipid homeostasis
oxidation-reduction process
ascaroside biosynthetic process
acyl-CoA oxidase activity
fatty acid binding
oxidoreductase activity, acting on the CH-CH group of donors
flavin adenine dinucleotide binding
FAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0