This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02423521_g1
See other KPC-1 GE Assays ›
Gene Symbol:
kpc-1
Gene Name:
Furin-like protease kpc-1
Gene Aliases:
CELE_F11A6.1
Chromosome Location:
Chr.I: 11675945 - 11680975 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02423521_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: kpc-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_060573.6 NP_492974.2 6 - 7 1987 73
NR_132618.1 5 - 6 2355 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
gene expression
protein processing
peptide hormone processing
insulin processing
zymogen activation
protein localization to endosome
positive regulation of synaptic growth at neuromuscular junction
dendrite morphogenesis
regulation of dendrite morphogenesis
positive regulation of dendrite morphogenesis
dendrite self-avoidance
positive regulation of dendrite development
regulation of dendrite extension
positive regulation of dendrite extension
negative regulation of dauer entry
negative regulation of protein localization to cell surface
regulation of neuron migration
serine-type endopeptidase activity
receptor binding
peptidase activity
serine-type peptidase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0