This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02422387_g1
See other WNK-1 GE Assays ›
Gene Symbol:
wnk-1
Gene Name:
protein-coding
Gene Aliases:
CELE_C46C2.1
Chromosome Location:
Chr.IV: 9200102 - 9212242 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02422387_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: wnk-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001268439.4 NP_001255368.1 3 - 4 477 91
NM_001268440.3 NP_001255369.1 2 - 3 156 91
NM_001268441.3 NP_001255370.1 3 - 4 485 91
NM_001268442.4 NP_001255371.1 2 - 3 156 91
NM_001268443.3 NP_001255372.1 2 - 3 156 91
NM_001268444.3 NP_001255373.1 2 - 3 156 91
NM_001307103.3 NP_001294032.1 3 - 4 481 91
NM_001307104.3 NP_001294033.1 2 - 3 156 91
NM_001307106.4 NP_001294035.1 2 - 3 156 91
NM_001307107.3 NP_001294036.1 3 - 4 477 91
NM_001307108.3 NP_001294037.1 2 - 3 156 91
NM_001307109.4 NP_001294038.1 3 - 4 473 91
NM_001380399.1 NP_001368454.1 3 - 4 477 91
NM_001380400.1 NP_001368092.1 3 - 4 477 91
NM_069202.6 NP_501603.3 3 - 4 471 91
NM_182275.7 NP_872075.2 3 - 4 473 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
protein phosphorylation
hyperosmotic response
multicellular organism development
negative regulation of sodium ion transport
phosphorylation
positive regulation of ion transmembrane transporter activity
intracellular signal transduction
ion homeostasis
multicellular organismal water homeostasis
positive regulation of potassium ion import
positive regulation of sodium ion transmembrane transporter activity
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
chloride channel inhibitor activity
potassium channel inhibitor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0