This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02422386_g1
See other WNK-1 GE Assays ›
Gene Symbol:
wnk-1
Gene Name:
protein-coding
Gene Aliases:
CELE_C46C2.1
Chromosome Location:
Chr.IV: 9200102 - 9212242 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02422386_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: wnk-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001268439.4 NP_001255368.1 2 - 3 410 99
NM_001268440.3 NP_001255369.1 1 - 2 89 99
NM_001268441.3 NP_001255370.1 2 - 3 418 99
NM_001268442.4 NP_001255371.1 1 - 2 89 99
NM_001268443.3 NP_001255372.1 1 - 2 89 99
NM_001268444.3 NP_001255373.1 1 - 2 89 99
NM_001307103.3 NP_001294032.1 2 - 3 414 99
NM_001307104.3 NP_001294033.1 1 - 2 89 99
NM_001307106.4 NP_001294035.1 1 - 2 89 99
NM_001307107.3 NP_001294036.1 2 - 3 410 99
NM_001307108.3 NP_001294037.1 1 - 2 89 99
NM_001307109.4 NP_001294038.1 2 - 3 406 99
NM_001380399.1 NP_001368454.1 2 - 3 410 99
NM_001380400.1 NP_001368092.1 2 - 3 410 99
NM_069202.6 NP_501603.3 2 - 3 404 99
NM_182275.7 NP_872075.2 2 - 3 406 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
protein phosphorylation
hyperosmotic response
multicellular organism development
negative regulation of sodium ion transport
phosphorylation
positive regulation of ion transmembrane transporter activity
intracellular signal transduction
ion homeostasis
multicellular organismal water homeostasis
positive regulation of potassium ion import
positive regulation of sodium ion transmembrane transporter activity
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
chloride channel inhibitor activity
potassium channel inhibitor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0