This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02421676_m1
See other DIN-1 GE Assays ›
Gene Symbol:
din-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F07A11.6
Chromosome Location:
Chr.II: 11614624 - 11633611 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02421676_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: din-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267377.3 NP_001254306.1 12 - 13 5475 136
NM_001381614.1 NP_001367110.1 18 - 19 6631 136
NM_064083.6 NP_496484.3 16 - 17 6089 136
NM_064084.8 NP_496485.2 17 - 18 6679 136
NM_182101.5 NP_871901.1 18 - 19 6609 136

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
regulation of transcription, DNA-templated
transcription regulatory region sequence-specific DNA binding
nucleic acid binding
RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0