This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ce02421139_g1
See other F49E2.5 GE Assays ›
Gene Symbol:
F49E2.5
Gene Name:
protein-coding
Gene Aliases:
CELE_F49E2.5
Chromosome Location:
Chr.X: 9543652 - 9560804 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID
Ce02421139_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
374.00
Your Price
Online offer:
374.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: F49E2.5
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001029523.5
NP_001024694.1
3 - 4
396
108
NM_001029524.6
NP_001024695.1
3 - 4
408
108
NM_001373430.3
NP_001360758.1
3 - 4
392
108
NM_001373432.4
NP_001360760.1
3 - 4
389
108
NM_001373433.2
NP_001360761.1
3 - 4
392
108
NM_001381054.2
NP_001368747.1
3 - 4
347
108
NM_001381055.1
NP_001366703.1
2 - 3
198
108
NM_001381056.1
NP_001366700.1
2 - 3
198
108
NM_001381057.1
NP_001367097.1
2 - 3
198
108
NM_001381058.1
NP_001368748.1
2 - 3
198
108
NM_001381059.1
NP_001368749.1
2 - 3
198
108
NM_001381060.1
NP_001368750.1
2 - 3
198
108
NM_001381061.2
NP_001368751.1
2 - 3
198
108
NM_001381062.1
NP_001368752.1
2 - 3
198
108
NM_001381063.1
NP_001368753.1
2 - 3
198
108
NM_001381064.1
NP_001366704.1
2 - 3
254
108
NM_001381065.1
NP_001368754.1
2 - 3
198
108
NM_001381066.1
NP_001368755.1
2 - 3
254
108
NM_001381067.1
NP_001367224.1
2 - 3
198
108
NM_001381069.1
NP_001367019.1
2 - 3
198
108
NM_001381070.1
NP_001366701.1
2 - 3
198
108
NM_001383624.1
2 - 3
248
108
NM_001383625.1
3 - 4
396
108
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Detect Genomic DNA
Caenorhabditis elegans
Probe spans exons
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0