This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02421121_m1
See other AJM-1 GE Assays ›
Gene Symbol:
ajm-1
Gene Name:
protein-coding
Gene Aliases:
CELE_C25A11.4
Chromosome Location:
Chr.X: 9082946 - 9113619 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02421121_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ajm-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001330908.3 NP_001317889.1 16 - 17 3929 57
NM_001330909.3 NP_001317890.1 16 - 17 3704 57
NM_001330910.3 NP_001317891.1 14 - 15 3572 57
NM_001330911.3 NP_001317888.1 13 - 14 3458 57
NM_001330912.3 NP_001317897.1 12 - 13 3142 57
NM_001330913.3 NP_001317898.1 11 - 12 2897 57
NM_001330914.3 NP_001317899.1 10 - 11 2825 57
NM_001330915.3 NP_001317736.1 8 - 9 2528 57
NM_001381045.1 NP_001368606.1 22 - 23 4712 57
NM_001381046.2 NP_001367383.1 21 - 22 4597 57
NM_001381048.1 NP_001367381.1 22 - 23 4749 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

embryo development ending in birth or egg hatching
cell-cell junction organization
protein binding
cytoskeletal protein binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0