This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02421057_m1
See other SLO-1 GE Assays ›
Gene Symbol:
slo-1
Gene Name:
protein-coding
Gene Aliases:
CELE_Y51A2D.19
Chromosome Location:
Chr.V: 18492261 - 18508212 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02421057_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slo-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029088.2 NP_001024259.1 11 - 12 2081 65
NM_001029089.3 NP_001024260.1 11 - 12 2087 65
NM_001269876.3 NP_001256805.1 11 - 12 2042 65
NM_001269877.3 NP_001256806.1 11 - 12 2042 65
GenBank mRNA AF431891.1 - 11 - 12 2042 65
AF431892.1 - 11 - 12 2042 65
JQ606810.1 - 11 - 12 2042 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ion transport
potassium ion transport
ion transmembrane transport
regulation of ion transmembrane transport
locomotion
response to drug
pharyngeal pumping
sarcomere organization
regulation of neurotransmitter secretion
behavioral response to ethanol
modulation of synaptic transmission
transmembrane transport
potassium ion transmembrane transport
ion channel activity
voltage-gated ion channel activity
voltage-gated potassium channel activity
potassium channel activity
protein binding
calcium-activated potassium channel activity
large conductance calcium-activated potassium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0