This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02420752_g1
See other MADD-3 GE Assays ›
Gene Symbol:
madd-3
Gene Name:
protein-coding
Gene Aliases:
CELE_E02H4.3
Chromosome Location:
Chr.X: 14284752 - 14296660 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02420752_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: madd-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001381143.1 NP_001368709.1 1 - 2 1538 112
NM_171799.8 NP_741928.1 1 - 2 3169 112

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
peptidyl-tyrosine phosphorylation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein tyrosine kinase activity
ATP binding
kinase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0