This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02420745_g1
See other MADD-3 GE Assays ›
Gene Symbol:
madd-3
Gene Name:
protein-coding
Gene Aliases:
CELE_E02H4.3
Chromosome Location:
Chr.X: 14284752 - 14296660 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02420745_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: madd-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001368549.3 NP_001355453.1 4 - 5 771 82
NM_001368550.3 NP_001355370.1 4 - 5 771 82
NM_001375168.2 NP_001362073.1 4 - 5 768 82
NM_001381143.1 NP_001368709.1 2 - 3 1651 82
NM_171799.8 NP_741928.1 2 - 3 3282 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001375168.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
peptidyl-tyrosine phosphorylation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein tyrosine kinase activity
ATP binding
kinase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0