This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02420595_g1
See other SMA-9 GE Assays ›
Gene Symbol:
sma-9
Gene Name:
protein-coding
Gene Aliases:
CELE_T05A10.1
Chromosome Location:
Chr.X: 10769282 - 10782973 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02420595_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sma-9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001375119.3 NP_001361905.1 16 - 17 4830 62
NM_001375120.2 NP_001361887.1 17 - 18 5277 62
NM_001375121.3 NP_001361888.1 17 - 18 5277 62
NM_001375122.2 NP_001361889.1 17 - 18 5277 62
NM_001375123.1 NP_001361886.1 17 - 18 5277 62
NM_001375124.1 NP_001361885.1 18 - 19 5358 62
NM_001375125.2 NP_001361884.1 16 - 17 4824 62
NM_001377620.1 NP_001364832.1 17 - 18 4953 62
NM_001383639.1 17 - 18 4955 62
NM_171771.6 NP_741896.2 15 - 16 4749 62
GenBank mRNA AY390550.1 - 7 - 8 1118 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transforming growth factor beta receptor signaling pathway
positive regulation of multicellular organism growth
regulation of mesodermal cell fate specification
nematode male tail tip morphogenesis
positive regulation of mesodermal cell fate specification
negative regulation of nucleic acid-templated transcription
positive regulation of nucleic acid-templated transcription
nucleic acid binding
transcription coactivator activity
transcription corepressor activity
zinc ion binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0