This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02420351_g1
See other F48E3.8 GE Assays ›
Gene Symbol:
F48E3.8
Gene Name:
protein-coding
Gene Aliases:
CELE_F48E3.8
Chromosome Location:
Chr.X: 7507877 - 7519669 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02420351_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: F48E3.8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001373360.2 NP_001360643.1 15 - 16 2342 77
NM_001373361.3 NP_001360755.1 11 - 12 1691 77
NM_001373362.3 NP_001360044.1 11 - 12 1691 77
NM_001373363.2 NP_001360754.1 12 - 13 1814 77
NM_001373364.2 NP_001360043.1 12 - 13 1814 77
NM_001373365.3 NP_001360756.1 12 - 13 1814 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
catalytic activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0