This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02419736_g1
See other UNC-2 GE Assays ›
Gene Symbol:
unc-2
Gene Name:
protein-coding
Gene Aliases:
CELE_T02C5.5
Chromosome Location:
Chr.X: 2710920 - 2763526 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02419736_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: unc-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001348588.4 NP_001335561.1 14 - 15 2587 132
NM_001348589.4 NP_001335563.1 14 - 15 2587 132
NM_001348590.4 NP_001335560.1 14 - 15 2587 132
NM_001348591.4 NP_001335562.1 14 - 15 2587 132
NM_001348592.5 NP_001335571.1 14 - 15 2782 132
NM_001348593.4 NP_001335573.1 14 - 15 2782 132
NM_001348594.5 NP_001335570.1 14 - 15 2782 132
NM_001348595.4 NP_001335572.1 14 - 15 2782 132
NM_001348596.5 NP_001335516.1 12 - 13 2314 132
NM_001348597.2 NP_001335515.1 12 - 13 2314 132
NM_001368494.4 NP_001355464.1 20 - 21 3148 132
NM_001368495.4 NP_001355463.1 18 - 19 2992 132
NM_001368496.4 NP_001355462.1 18 - 19 3016 132
NM_001368497.4 NP_001355494.1 14 - 15 2915 132
NM_171638.3 NP_741732.2 14 - 15 2772 132
GenBank mRNA AY264781.1 - 14 - 15 2772 132

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of neurotransmitter levels
ion transport
calcium ion transport
chemical synaptic transmission
locomotory behavior
response to heat
negative regulation of transforming growth factor beta receptor signaling pathway
regulation of ion transmembrane transport
determination of left/right asymmetry in nervous system
response to starvation
transmembrane transport
calcium ion import
calcium ion transmembrane transport
ion channel activity
voltage-gated ion channel activity
voltage-gated calcium channel activity
calcium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0