This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02419370_g1
See other SLO-1 GE Assays ›
Gene Symbol:
slo-1
Gene Name:
protein-coding
Gene Aliases:
CELE_Y51A2D.19
Chromosome Location:
Chr.V: 18492261 - 18508212 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02419370_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slo-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029088.2 NP_001024259.1 19 - 20 3398 103
NM_001029089.3 NP_001024260.1 18 - 19 3338 103
NM_001029090.5 NP_001024261.1 19 - 20 3377 103
NM_001269872.3 NP_001256801.1 20 - 21 3377 103
NM_001269873.3 NP_001256802.1 20 - 21 3419 103
NM_001269874.3 NP_001256803.1 19 - 20 3353 103
NM_001269875.4 NP_001256804.1 20 - 21 3443 103
NM_001269876.3 NP_001256805.1 19 - 20 3359 103
NM_001269877.3 NP_001256806.1 18 - 19 3293 103
NM_001269878.3 NP_001256807.1 20 - 21 3419 103
NM_001269879.4 NP_001256808.1 19 - 20 3398 103
NM_001269880.3 NP_001256809.1 20 - 21 3398 103
NM_001269881.3 NP_001256810.1 19 - 20 3332 103
GenBank mRNA AF431891.1 - 19 - 20 3359 103
AF431892.1 - 18 - 19 3293 103
AF431893.1 - 19 - 20 3332 103
JQ582463.1 - 20 - 21 3419 103
JQ606809.1 - 20 - 21 3398 103
JQ606810.1 - 18 - 19 3293 103
JQ606812.1 - 19 - 20 3332 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ion transport
potassium ion transport
ion transmembrane transport
regulation of ion transmembrane transport
locomotion
response to drug
pharyngeal pumping
sarcomere organization
regulation of neurotransmitter secretion
behavioral response to ethanol
modulation of synaptic transmission
transmembrane transport
potassium ion transmembrane transport
ion channel activity
voltage-gated ion channel activity
voltage-gated potassium channel activity
potassium channel activity
protein binding
calcium-activated potassium channel activity
large conductance calcium-activated potassium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0