This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02419365_m1
See other SLO-1 GE Assays ›
Gene Symbol:
slo-1
Gene Name:
protein-coding
Gene Aliases:
CELE_Y51A2D.19
Chromosome Location:
Chr.V: 18492261 - 18508212 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02419365_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slo-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029088.2 NP_001024259.1 14 - 15 2326 117
NM_001029089.3 NP_001024260.1 13 - 14 2266 117
NM_001029090.5 NP_001024261.1 14 - 15 2305 117
NM_001269872.3 NP_001256801.1 15 - 16 2305 117
NM_001269873.3 NP_001256802.1 15 - 16 2347 117
NM_001269874.3 NP_001256803.1 14 - 15 2281 117
NM_001269875.4 NP_001256804.1 15 - 16 2371 117
NM_001269876.3 NP_001256805.1 14 - 15 2287 117
NM_001269877.3 NP_001256806.1 13 - 14 2221 117
NM_001269878.3 NP_001256807.1 15 - 16 2347 117
NM_001269879.4 NP_001256808.1 14 - 15 2326 117
NM_001269880.3 NP_001256809.1 15 - 16 2326 117
NM_001269881.3 NP_001256810.1 14 - 15 2260 117
GenBank mRNA AF431891.1 - 14 - 15 2287 117
AF431892.1 - 13 - 14 2221 117
AF431893.1 - 14 - 15 2260 117
JQ582463.1 - 15 - 16 2347 117
JQ606809.1 - 15 - 16 2326 117
JQ606810.1 - 13 - 14 2221 117
JQ606812.1 - 14 - 15 2260 117

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ion transport
potassium ion transport
ion transmembrane transport
regulation of ion transmembrane transport
locomotion
response to drug
pharyngeal pumping
sarcomere organization
regulation of neurotransmitter secretion
behavioral response to ethanol
modulation of synaptic transmission
transmembrane transport
potassium ion transmembrane transport
ion channel activity
voltage-gated ion channel activity
voltage-gated potassium channel activity
potassium channel activity
protein binding
calcium-activated potassium channel activity
large conductance calcium-activated potassium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0