This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02418605_g1
See other RTEL-1 GE Assays ›
Gene Symbol:
rtel-1 F29C6.1
Gene Name:
Regulator of telomere elongation helicase 1 homolog
protein-coding
Gene Aliases:
CELE_F25H2.13
CELE_F29C6.1
Chromosome Location:
Chr.I: 10571954 - 10577135 on Build WBcel235
Chromosome Location:
Chr.I: 10572023 - 10580960 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02418605_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rtel-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_060368.6 NP_492769.1 4 - 5 1066 176
Gene Symbol: F29C6.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001381203.1 NP_001367064.1 4 - 5 1058 176

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001381203.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

nucleobase-containing compound metabolic process
DNA repair
cellular response to DNA damage stimulus
regulation of double-strand break repair via homologous recombination
DNA duplex unwinding
negative regulation of DNA recombination
telomeric loop disassembly
negative regulation of t-circle formation
nucleotide binding
nucleic acid binding
DNA binding
DNA helicase activity
helicase activity
ATP binding
hydrolase activity
hydrolase activity, acting on acid anhydrides, in phosphorus-containing anhydrides
metal ion binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding
DNA polymerase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0