This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02418408_g1
See other AIR-2 GE Assays ›
Gene Symbol:
air-2
Gene Name:
Aurora/IPL1-related protein kinase 2
Gene Aliases:
CELE_B0207.4
Chromosome Location:
Chr.I: 5937195 - 5963872 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02418408_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: air-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_059313.8 NP_491714.1 2 - 3 148 94
GenBank mRNA AF071207.1 - 1 - 2 86 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
protein phosphorylation
cell cycle
mitotic spindle organization
chromosome segregation
multicellular organism development
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
chromosome condensation
regulation of actomyosin contractile ring contraction
regulation of cytokinesis
histone-serine phosphorylation
establishment of protein localization
protein autophosphorylation
mitotic spindle midzone assembly
meiotic spindle midzone assembly
cell division
meiotic cell cycle
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
kinesin binding
histone serine kinase activity
ATPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0