This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02418366_g1
See other STAM-1 GE Assays ›
Gene Symbol:
stam-1
Gene Name:
protein-coding
Gene Aliases:
CELE_C34G6.7
Chromosome Location:
Chr.I: 5902085 - 5904689 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02418366_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: stam-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_059309.8 NP_491710.2 4 - 5 552 78
NM_182013.8 NP_871813.1 3 - 4 496 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
multicellular organism development
endosome to lysosome transport
protein transport
response to hermaphrodite contact
protein targeting to vacuole involved in ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
negative regulation of MAPK cascade
ephrin receptor signaling pathway
protein localization to nonmotile primary cilium
protein binding
protein C-terminus binding
ubiquitin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0