This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02417501_g1
See other LET-363 GE Assays ›
Gene Symbol:
let-363
Gene Name:
protein-coding
Gene Aliases:
CELE_B0261.2
Chromosome Location:
Chr.I: 5245743 - 5263426 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02417501_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: let-363
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_059148.5 NP_491549.2 28 - 29 7038 86
NM_059151.7 NP_491552.2 28 - 29 6991 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

box C/D snoRNP assembly
nematode larval development
regulation of translation
protein phosphorylation
determination of adult lifespan
negative regulation of autophagy
positive regulation of gene expression
negative regulation of gene expression
negative regulation of macroautophagy
phosphorylation
TOR signaling
TORC1 signaling
positive regulation of translation
mesendoderm development
response to sodium arsenite
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
macromolecular complex binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0