This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02415948_g1
See other UBR-1 GE Assays ›
Gene Symbol:
ubr-1
Gene Name:
E3 ubiquitin-protein ligase ubr-1
Gene Aliases:
CELE_C32E8.11
Chromosome Location:
Chr.I: 3807456 - 3816809 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02415948_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ubr-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_058830.6 NP_491231.2 7 - 8 2739 137

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
protein catabolic process
regulation of backward locomotion
ubiquitin-dependent protein catabolic process via the N-end rule pathway
negative regulation of glutamate metabolic process
zinc ion binding
transferase activity
ubiquitin conjugating enzyme binding
metal ion binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0