This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02415896_g1
See other ASIC-1 GE Assays ›
Gene Symbol:
asic-1 C32E8.6
Gene Name:
Degenerin-like protein asic-1
protein-coding
Gene Aliases:
CELE_ZK770.1
CELE_C32E8.6
Chromosome Location:
Chr.I: 3769672 - 3774005 on Build WBcel235
Chromosome Location:
Chr.I: 3769662 - 3777468 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02415896_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: asic-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_058813.5 NP_491214.3 2 - 3 325 145
Gene Symbol: C32E8.6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001384008.1 2 - 3 335 145

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001384008.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ion transport
sodium ion transport
positive regulation of dopamine secretion
sodium ion transmembrane transport
protein homotrimerization
cellular response to pH
ion channel activity
sodium channel activity
ligand-gated sodium channel activity
acid-sensing ion channel activity
catalytic activity
CoA-ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0