This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02415516_g1
See other W05F2.4 GE Assays ›
Gene Symbol:
W05F2.4
Gene Name:
protein-coding
Gene Aliases:
CELE_W05F2.4
Chromosome Location:
Chr.I: 3374776 - 3407189 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02415516_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: W05F2.4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037894.4 NP_001032983.2 34 - 35 5536 64
NM_001037895.5 NP_001032984.2 10 - 11 2166 64
NM_001262432.3 NP_001249361.1 12 - 13 2369 64
NM_001262433.4 NP_001249362.1 36 - 37 5762 64
NM_001381630.2 NP_001367784.1 11 - 12 2125 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0