This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02414646_g1
See other ABT-2 GE Assays ›
Gene Symbol:
abt-2
Gene Name:
protein-coding
Gene Aliases:
CELE_F12B6.1
Chromosome Location:
Chr.I: 2255921 - 2272866 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02414646_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: abt-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001381367.1 NP_001368466.1 25 - 26 4352 94
NM_058548.7 NP_490949.3 25 - 26 4463 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid transport
transmembrane transport
nucleotide binding
lipid transporter activity
ATP binding
ATPase activity
ATPase activity, coupled to transmembrane movement of substances

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0