This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02414325_g1
See other BRE-4 GE Assays ›
Gene Symbol:
bre-4
Gene Name:
protein-coding
Gene Aliases:
CELE_Y73E7A.7
Chromosome Location:
Chr.I: 1621314 - 1628486 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02414325_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: bre-4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001383737.1 4 - 5 689 65
NM_058471.2 NP_490872.1 4 - 5 694 65
GenBank mRNA AY130767.1 - 3 - 4 596 65
AY533306.1 - 3 - 4 596 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
protein glycosylation
response to toxic substance
positive regulation of Notch signaling pathway
acetylgalactosaminyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
beta-1,4-N-acetylgalactosaminyltransferase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0