This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02414303_m1
See other SRC-1 GE Assays ›
Gene Symbol:
src-1
Gene Name:
protein-coding
Gene Aliases:
CELE_Y92H12A.1
Chromosome Location:
Chr.I: 1567602 - 1580208 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02414303_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: src-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001306492.4 NP_001293421.1 1 - 2 247 64
NM_001377652.2 NP_001364829.1 2 - 3 427 64
NM_001377653.2 NP_001364774.1 2 - 3 422 64
GenBank mRNA AF419171.1 - 3 - 4 512 64
AF475094.1 - 2 - 3 415 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

establishment of mitotic spindle orientation
endodermal cell fate specification
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
anterior/posterior axis specification, embryo
embryo development ending in birth or egg hatching
phosphorylation
cell differentiation
gonad morphogenesis
netrin-activated signaling pathway
peptidyl-tyrosine autophosphorylation
positive regulation of locomotion
positive regulation of embryonic development
regulation of cell proliferation
innate immune response
protein autophosphorylation
embryonic digestive tract morphogenesis
positive regulation of peptidyl-tyrosine phosphorylation
left/right axis specification
commissural neuron axon guidance
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
kinase activity
transferase activity
metal ion binding
netrin receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0