This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02413869_g1
See other PIF-1 GE Assays ›
Gene Symbol:
pif-1
Gene Name:
protein-coding
Gene Aliases:
CELE_Y18H1A.6
Chromosome Location:
Chr.I: 719929 - 731078 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02413869_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pif-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001306245.3 NP_001293174.1 2 - 3 249 67
NM_058373.6 NP_490774.2 4 - 5 716 67
GenBank mRNA AB015041.1 - 4 - 5 573 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial genome maintenance
telomere maintenance
DNA replication
DNA repair
DNA recombination
cellular response to DNA damage stimulus
DNA duplex unwinding
nucleotide binding
DNA binding
DNA helicase activity
helicase activity
ATP binding
hydrolase activity
5'-3' DNA helicase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0