This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02413808_g1
See other HUM-7 GE Assays ›
Gene Symbol:
hum-7
Gene Name:
protein-coding
Gene Aliases:
CELE_F56A6.2
Chromosome Location:
Chr.I: 585807 - 607571 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02413808_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hum-7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261950.4 NP_001248879.1 8 - 9 1492 100
NM_001261951.3 NP_001248880.1 8 - 9 1492 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
signal transduction
vesicle transport along actin filament
actin-myosin filament sliding
intracellular signal transduction
microfilament motor activity
nucleotide binding
motor activity
actin binding
calmodulin binding
ATP binding
actin-dependent ATPase activity
metal ion binding
actin filament binding
plus-end directed microfilament motor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0