This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02413240_m1
See other PDPR-1 GE Assays ›
Gene Symbol:
pdpr-1 madf-10
Gene Name:
Pyruvate Dehydrogenase Phosphatase Regulatory subunit
MADF domain-containing protein
Gene Aliases:
CELE_Y106G6H.5
CELE_Y106G6H.4
Chromosome Location:
Chr.I: 10438075 - 10443563 on Build WBcel235
Chromosome Location:
Chr.I: 10436895 - 10439443 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02413240_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pdpr-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_060329.5 NP_492730.2 10 - 11 2407 142
Gene Symbol: madf-10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_060328.3 NP_492729.1 1 - 2 304 142

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_060328.3 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
regulation of transcription from RNA polymerase II promoter
oxidoreductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0