This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02412396_g1
See other ATE-1 GE Assays ›
Gene Symbol:
ate-1
Gene Name:
protein-coding
Gene Aliases:
CELE_K07A1.9
Chromosome Location:
Chr.I: 9611813 - 9615084 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02412396_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ate-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001348617.2 NP_001335527.1 5 - 6 564 90
NM_001348618.3 NP_001335528.1 3 - 4 338 90
NM_001348619.3 NP_001335529.1 3 - 4 356 90
NM_001373508.1 NP_001359652.1 6 - 7 632 90
NM_060148.7 NP_492549.2 5 - 6 570 90
NM_182029.4 NP_871829.1 4 - 5 484 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein arginylation
arginyltransferase activity
transferase activity
transferase activity, transferring acyl groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0