This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02411941_g1
See other C45G3.3 GE Assays ›
Gene Symbol:
C45G3.3 gip-2
Gene Name:
BPL/LPL catalytic domain-containing protein
Gamma-tubulin interacting protein 2
Gene Aliases:
CELE_C45G3.3
CELE_C45G3.5
Chromosome Location:
Chr.I: 9226905 - 9232103 on Build WBcel235
Chromosome Location:
Chr.I: 9228163 - 9232101 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02411941_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C45G3.3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001381008.1 NP_001366919.1 6 - 7 1268 78
Gene Symbol: gip-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025873.6 NP_001021044.1 2 - 3 203 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein lipoylation
lipoate-protein ligase activity
lipoyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0