This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02411442_g1
See other UBR-5 GE Assays ›
Gene Symbol:
ubr-5
Gene Name:
UBR E3 ubiquitin ligase homolog
Gene Aliases:
CELE_F36A2.13
Chromosome Location:
Chr.I: 8828010 - 8838719 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02411442_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ubr-5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_059988.5 NP_492389.1 6 - 7 910 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
cellular response to DNA damage stimulus
negative regulation of histone H2A K63-linked ubiquitination
regulation of double-strand break repair
ubiquitin-protein transferase activity
zinc ion binding
transferase activity
ubiquitin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0