This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02411015_g1
See other R05D11.9 GE Assays ›
Gene Symbol:
R05D11.9 paxt-1
Gene Name:
Uncharacterized protein
Partner of xrn-2 protein 1
Gene Aliases:
CELE_R05D11.9
CELE_R05D11.6
Chromosome Location:
Chr.I: 8593027 - 8596225 on Build WBcel235
Chromosome Location:
Chr.I: 8593025 - 8598558 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02411015_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: R05D11.9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_059923.10 NP_492324.2 2 - 3 336 147
Gene Symbol: paxt-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001383484.1 2 - 3 338 147

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001383484.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

sphingomyelin catabolic process
glycerophospholipid catabolic process
ceramide biosynthetic process
mRNA processing
sphingomyelin phosphodiesterase D activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0