This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02410330_g1
See other CHE-3 GE Assays ›
Gene Symbol:
che-3
Gene Name:
Cytoplasmic dynein 2 heavy chain 1
Gene Aliases:
CELE_F18C12.1
Chromosome Location:
Chr.I: 8062007 - 8081671 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02410330_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: che-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_059820.4 NP_492221.2 32 - 33 8179 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

microtubule-based movement
multicellular organism development
chemosensory behavior
protein localization
cell projection organization
negative regulation of transforming growth factor beta receptor signaling pathway
intraciliary retrograde transport
dauer entry
cilium morphogenesis
positive regulation of dauer larval development
non-motile cilium assembly
nucleotide binding
motor activity
microtubule motor activity
ATP binding
ATP-dependent microtubule motor activity, minus-end-directed
dynein intermediate chain binding
dynein light intermediate chain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0