This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02409900_g1
See other GLD-1 GE Assays ›
Gene Symbol:
gld-1 T23G11.4
Gene Name:
Female germline-specific tumor suppressor gld-1
Uncharacterized protein
Gene Aliases:
CELE_T23G11.3
CELE_T23G11.4
Chromosome Location:
Chr.I: 7695661 - 7698227 on Build WBcel235
Chromosome Location:
Chr.I: 7694320 - 7698229 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02409900_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gld-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_059742.7 NP_492143.1 6 - 7 945 67
Gene Symbol: T23G11.4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001383384.1 6 - 7 1529 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
regulation of translation
multicellular organism development
regulation of gene expression
negative regulation of gene expression
negative regulation of translation
cell differentiation
oocyte fate determination
oogenesis
meiotic cell cycle
positive regulation of oocyte development
regulation of germ cell proliferation
positive regulation of female gonad development
negative regulation of nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
regulation of mRNA splicing, via spliceosome
nucleic acid binding
RNA binding
single-stranded RNA binding
mRNA binding
mRNA 3'-UTR binding
protein binding
mRNA 5'-UTR binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0