This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02409042_g1
See other PDI-3 GE Assays ›
Gene Symbol:
pdi-3
Gene Name:
Protein disulfide-isomerase
Gene Aliases:
CELE_H06O01.1
Chromosome Location:
Chr.I: 7015912 - 7020790 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02409042_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pdi-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_059594.8 NP_491995.1 6 - 7 1227 72
GenBank mRNA AB072567.1 - 6 - 7 1229 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein folding
regulation of cell shape
body morphogenesis
peptide cross-linking
oviposition
response to endoplasmic reticulum stress
collagen and cuticulin-based cuticle development
positive regulation of embryonic development
nematode male tail tip morphogenesis
cell redox homeostasis
protein deglutathionylation
nematode male tail mating organ morphogenesis
protein disulfide isomerase activity
protein-glutamine gamma-glutamyltransferase activity
isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0