This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02408176_g1
See other GLD-2 GE Assays ›
Gene Symbol:
gld-2
Gene Name:
protein-coding
Gene Aliases:
CELE_ZC308.1
Chromosome Location:
Chr.I: 6369748 - 6386194 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02408176_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gld-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001026673.3 NP_001021844.1 4 - 5 1402 146
NM_001313548.3 NP_001300477.1 8 - 9 1271 146
NM_001313549.3 NP_001300478.1 6 - 7 992 146
NM_059440.5 NP_491841.2 7 - 8 1875 146
NM_059441.5 NP_491842.2 15 - 16 2285 146
GenBank mRNA AY125085.1 - 14 - 15 2133 146

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mRNA polyadenylation
mRNA processing
nucleus organization
multicellular organism development
embryo development ending in birth or egg hatching
RNA polyadenylation
regulation of cell division
meiotic cell cycle
positive regulation of meiosis I
nucleotide binding
polynucleotide adenylyltransferase activity
protein binding
ATP binding
transferase activity
nucleotidyltransferase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0